siRNA Details
| Gene Symbol: Cetn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191842.1 | NP_001178771.1 | 3 | 331 | ||
| GenBank mRNA | AF335277.2 | - | 2 | 221 | ||
| FQ234873.1 | - | 3 | 264 | |||
| Gene Symbol: LOC100912538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749163.3 | XP_003749211.1 | 3 | 1439 | ||

Cetn3
Genomic Map