siRNA Details
| Gene Symbol: Rbm14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133388.1 | NP_596879.1 | 2 | 1077 | ||
| GenBank mRNA | AF327567.1 | - | 1 | 53 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm14| Gene Symbol: Rbm14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133388.1 | NP_596879.1 | 2 | 1077 | ||
| GenBank mRNA | AF327567.1 | - | 1 | 53 | ||