siRNA Details
Gene Symbol: Rbm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133388.1 | NP_596879.1 | 2 | 1367 | ||
GenBank mRNA | AF327567.1 | - | 1 | 343 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133388.1 | NP_596879.1 | 2 | 1367 | ||
GenBank mRNA | AF327567.1 | - | 1 | 343 |