siRNA Details
Gene Symbol: Pik3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133399.2 | NP_596890.2 | 16, 17 | 2493 | ||
GenBank mRNA | AF395897.1 | - | 4, 5 | 381 | ||
CR754046.1 | - | 3, 4 | 309 |
Gene Symbol: LOC685590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 2613 |
