siRNA Details
| Gene Symbol: Rap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133410.1 | NP_596901.1 | 1 | 371 | ||
| GenBank mRNA | AF386786.1 | - | 1 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rap2b| Gene Symbol: Rap2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133410.1 | NP_596901.1 | 1 | 371 | ||
| GenBank mRNA | AF386786.1 | - | 1 | 371 | ||