siRNA Details
Gene Symbol: Rap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133410.1 | NP_596901.1 | 1 | 483 | ||
GenBank mRNA | AF386786.1 | - | 1 | 483 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133410.1 | NP_596901.1 | 1 | 483 | ||
GenBank mRNA | AF386786.1 | - | 1 | 483 |