siRNA Details
| Gene Symbol: Ppp1r14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133425.3 | NP_596916.1 | 4 | 1082 | ||
| GenBank mRNA | AF407168.3 | - | 4 | 1082 | ||
| BC086978.1 | - | 4 | 555 | 7191557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r14c| Gene Symbol: Ppp1r14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133425.3 | NP_596916.1 | 4 | 1082 | ||
| GenBank mRNA | AF407168.3 | - | 4 | 1082 | ||
| BC086978.1 | - | 4 | 555 | 7191557 | ||