siRNA Details
| Gene Symbol: Zfp384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034830.1 | NP_001030002.1 | 10 | 1459 | ||
| NM_001034831.1 | NP_001030003.1 | 9 | 1275 | |||
| NM_133429.2 | NP_596920.2 | 10 | 1542 | |||
| XM_006237326.2 | XP_006237388.1 | 8 | 1320 | |||
| XM_006237327.2 | XP_006237389.1 | 11 | 1507 | |||
| XM_006237328.2 | XP_006237390.1 | 8 | 1255 | |||
| XM_006237329.2 | XP_006237391.1 | 8 | 1185 | |||
| GenBank mRNA | AB019281.1 | - | 10 | 1459 | ||
| AF216804.1 | - | 10 | 1542 | |||
| AF216805.1 | - | 8 | 1261 | |||
| AF216806.1 | - | 9 | 1275 | |||

Zfp384
Genomic Map