siRNA Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133519.4 | NP_598203.4 | 4 | 399 | ||
| GenBank mRNA | AF347935.1 | - | 4 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il11| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133519.4 | NP_598203.4 | 4 | 399 | ||
| GenBank mRNA | AF347935.1 | - | 4 | 416 | ||