siRNA Details
| Gene Symbol: Sstr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133522.1 | NP_598206.1 | 3 | 1054 | ||
| XM_006241966.2 | XP_006242028.1 | 2 | 1256 | |||
| GenBank mRNA | X63574.1 | - | 3 | 1054 | ||
More Information
Other Species Targeted
Set Membership: |
|

Sstr3
Genomic Map