siRNA Details
| Gene Symbol: Ccr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133532.2 | NP_598216.2 | 1 | 353 | ||
| XM_006243915.2 | XP_006243977.1 | 2 | 540 | |||
| GenBank mRNA | AF432872.2 | - | 1 | 353 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ccr4
Genomic Map