siRNA Details
| Gene Symbol: Hdgfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133549.2 | NP_598233.2 | 1 | 225 | ||
| GenBank mRNA | AF458586.1 | - | 1 | 195 | ||
| AY061636.1 | - | 1 | 207 | |||
| BC085707.1 | - | 1 | 225 | 7132556 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdgfl1| Gene Symbol: Hdgfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133549.2 | NP_598233.2 | 1 | 225 | ||
| GenBank mRNA | AF458586.1 | - | 1 | 195 | ||
| AY061636.1 | - | 1 | 207 | |||
| BC085707.1 | - | 1 | 225 | 7132556 | ||