siRNA Details
| Gene Symbol: Cst6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133566.1 | NP_598250.1 | 2 | 535 | ||
| GenBank mRNA | AY046415.1 | - | 2 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst6| Gene Symbol: Cst6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133566.1 | NP_598250.1 | 2 | 535 | ||
| GenBank mRNA | AY046415.1 | - | 2 | 535 | ||