siRNA Details
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133580.1 | NP_598264.1 | 3 | 230 | ||
GenBank mRNA | BC061984.1 | - | 4 | 538 | 5599180 | |
U18771.1 | - | 3 | 230 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133580.1 | NP_598264.1 | 3 | 230 | ||
GenBank mRNA | BC061984.1 | - | 4 | 538 | 5599180 | |
U18771.1 | - | 3 | 230 |