siRNA Details
| Gene Symbol: Wfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133581.1 | NP_598265.1 | 1 | 124 | ||
| XM_006255705.2 | XP_006255767.1 | 1 | 377 | |||
| GenBank mRNA | AF037272.1 | - | 1 | 124 | ||
| BC063152.1 | - | 1 | 127 | 6887007 | ||
| FQ213083.1 | - | 1 | 109 | |||
| FQ216345.1 | - | 1 | 118 | |||
| FQ216415.1 | - | 1 | 119 | |||
| FQ219846.1 | - | 1 | 119 | |||
| FQ220334.1 | - | 1 | 356 | |||
| FQ229404.1 | - | 1 | 109 | |||

Wfdc1
Genomic Map