siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133585.3 | NP_598269.3 | 13, 14 | 1439 | ||
| XM_006248497.1 | XP_006248559.1 | 15, 16 | 1703 | |||
| XM_006248498.1 | XP_006248560.1 | 14, 15 | 1649 | |||
| XM_006248499.1 | XP_006248561.1 | 14, 15 | 1592 | |||
| GenBank mRNA | AY333988.1 | - | 13, 14 | 1348 | ||
| AY510274.1 | - | 13, 14 | 1494 | |||
| BC111071.1 | - | 13, 14 | 1439 | 7324017 | ||

Opa1
Genomic Map