siRNA Details
| Gene Symbol: Rph3al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133591.2 | NP_598275.1 | 2 | 136 | ||
| XM_008767941.1 | XP_008766163.1 | 2 | 497 | |||
| XM_008767942.1 | XP_008766164.1 | 2 | 562 | |||
| XM_008767943.1 | XP_008766165.1 | 2 | 497 | |||
| XM_008767944.1 | XP_008766166.1 | 2 | 497 | |||
| GenBank mRNA | AF022774.1 | - | 1 | 62 | ||
| BC091126.1 | - | 2 | 136 | 7380477 | ||

Rph3al
Genomic Map