siRNA Details
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133610.2 | NP_598294.1 | 7, 8 | 1641 | ||
GenBank mRNA | AF185637.1 | - | 7, 8 | 1600 | ||
AJ250280.1 | - | 7, 8 | 1643 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133610.2 | NP_598294.1 | 7, 8 | 1641 | ||
GenBank mRNA | AF185637.1 | - | 7, 8 | 1600 | ||
AJ250280.1 | - | 7, 8 | 1643 |