siRNA Details
| Gene Symbol: Gpha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133619.2 | NP_598303.1 | 3 | 393 | ||
| XM_008760069.1 | XP_008758291.1 | 3 | 2429 | |||
| XM_008760070.1 | XP_008758292.1 | 3 | 2270 | |||
| GenBank mRNA | AF260741.1 | - | 3 | 393 | ||

Gpha2
Genomic Map