siRNA Details
| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134329.1 | NP_599156.1 | 8, 9 | 1158 | ||
| XM_008761493.1 | XP_008759715.1 | 9, 10 | 1545 | |||
| GenBank mRNA | X98746.1 | - | 8, 9 | 1158 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adh7| Gene Symbol: Adh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134329.1 | NP_599156.1 | 8, 9 | 1158 | ||
| XM_008761493.1 | XP_008759715.1 | 9, 10 | 1545 | |||
| GenBank mRNA | X98746.1 | - | 8, 9 | 1158 | ||