siRNA Details
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134329.1 | NP_599156.1 | 2 | 162 | ||
XM_008761493.1 | XP_008759715.1 | 3 | 549 | |||
GenBank mRNA | X98746.1 | - | 2 | 162 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134329.1 | NP_599156.1 | 2 | 162 | ||
XM_008761493.1 | XP_008759715.1 | 3 | 549 | |||
GenBank mRNA | X98746.1 | - | 2 | 162 |