siRNA Details
| Gene Symbol: Pou4f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134355.1 | NP_599182.1 | 2 | 1269 | ||
| XM_008772467.1 | XP_008770689.1 | 3 | 867 | |||
| GenBank mRNA | AF390076.1 | - | 1 | 69 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou4f2| Gene Symbol: Pou4f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134355.1 | NP_599182.1 | 2 | 1269 | ||
| XM_008772467.1 | XP_008770689.1 | 3 | 867 | |||
| GenBank mRNA | AF390076.1 | - | 1 | 69 | ||