siRNA Details
| Gene Symbol: Foxc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101680.1 | NP_001095150.1 | 1 | 811 | ||
| GenBank mRNA | L13193.1 | - | 1 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxc2| Gene Symbol: Foxc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101680.1 | NP_001095150.1 | 1 | 811 | ||
| GenBank mRNA | L13193.1 | - | 1 | 192 | ||