siRNA Details
| Gene Symbol: Cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134360.1 | NP_599187.1 | 2 | 123 | ||
| XM_006235511.2 | XP_006235573.1 | 2 | 185 | |||
| XM_008762463.1 | XP_008760685.1 | 2 | 186 | |||
| XM_008762464.1 | XP_008760686.1 | 2 | 184 | |||
| XM_008762465.1 | XP_008760687.1 | 2 | 185 | |||
| GenBank mRNA | AF241231.2 | - | 2 | 138 | ||
| BC097949.1 | - | 2 | 123 | 7387440 | ||
| FQ226992.1 | - | 2 | 106 | |||
| FQ231420.1 | - | 2 | 126 | |||

Cd40
Genomic Map