siRNA Details
| Gene Symbol: Cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134360.1 | NP_599187.1 | 5, 6 | 523 | ||
| XM_006235511.2 | XP_006235573.1 | 5, 6 | 585 | |||
| XM_008762464.1 | XP_008760686.1 | 5, 6 | 584 | |||
| GenBank mRNA | BC097949.1 | - | 5, 6 | 523 | 7387440 | |
| FQ226992.1 | - | 5, 6 | 506 | |||
| FQ231420.1 | - | 5, 6 | 526 | |||

Cd40
Genomic Map