siRNA Details
| Gene Symbol: Xcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134361.1 | NP_599188.1 | 3 | 204 | ||
| GenBank mRNA | U23377.1 | - | 3 | 204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xcl1| Gene Symbol: Xcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134361.1 | NP_599188.1 | 3 | 204 | ||
| GenBank mRNA | U23377.1 | - | 3 | 204 | ||