siRNA Details
| Gene Symbol: Rnf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134374.2 | NP_599201.1 | 4 | 1063 | ||
| GenBank mRNA | AB070897.1 | - | 4 | 1063 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf39| Gene Symbol: Rnf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134374.2 | NP_599201.1 | 4 | 1063 | ||
| GenBank mRNA | AB070897.1 | - | 4 | 1063 | ||