siRNA Details
| Gene Symbol: Clstn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134376.1 | NP_599203.1 | 14 | 2238 | ||
| GenBank mRNA | AJ431642.1 | - | 14 | 2238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clstn3| Gene Symbol: Clstn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134376.1 | NP_599203.1 | 14 | 2238 | ||
| GenBank mRNA | AJ431642.1 | - | 14 | 2238 | ||