siRNA Details
| Gene Symbol: UST4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134379.1 | NP_599206.1 | 7 | 1240 | ||
| GenBank mRNA | AJ132859.1 | - | 7 | 1240 | ||
| BC100065.1 | - | 1130 | 7373541 | |||
| FQ210050.1 | - | 1161 | ||||
| FQ218143.1 | - | 1300 | ||||

UST4r
Genomic Map