siRNA Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134384.1 | NP_599211.1 | 3 | 236 | ||
| GenBank mRNA | AB057408.1 | - | 3 | 236 | ||
| BC081729.1 | - | 3 | 253 | 7132055 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata19| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134384.1 | NP_599211.1 | 3 | 236 | ||
| GenBank mRNA | AB057408.1 | - | 3 | 236 | ||
| BC081729.1 | - | 3 | 253 | 7132055 | ||