siRNA Details
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134394.2 | NP_599221.1 | 2, 3 | 397 | ||
GenBank mRNA | AF175224.1 | - | 2, 3 | 400 | ||
BC078743.1 | - | 2, 3 | 409 | 7112937 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134394.2 | NP_599221.1 | 2, 3 | 397 | ||
GenBank mRNA | AF175224.1 | - | 2, 3 | 400 | ||
BC078743.1 | - | 2, 3 | 409 | 7112937 |