siRNA Details
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135169.2 | NP_001128641.1 | 5, 6 | 415 | ||
NM_134395.2 | NP_599222.2 | 4, 5 | 628 | |||
XM_006253777.2 | XP_006253839.1 | 5, 6 | 618 | |||
GenBank mRNA | AF083396.1 | - | 4 | 636 | ||
BC158854.1 | - | 4, 5 | 309 | 7936967 | ||
CV107072.1 | - | 1, 2 | 455 | |||
FQ211234.1 | - | 5, 6 | 405 | |||
FQ217516.1 | - | 5, 6 | 405 | |||
FQ218209.1 | - | 5, 6 | 411 | |||
FQ223586.1 | - | 5, 6 | 395 | |||
FQ223742.1 | - | 5, 6 | 405 | |||
FQ224495.1 | - | 5, 6 | 411 | |||
FQ225707.1 | - | 5, 6 | 385 | |||
FQ230083.1 | - | 5, 6 | 433 | |||
FQ230323.1 | - | 5, 6 | 414 | |||
FQ231364.1 | - | 5, 6 | 395 | |||
FQ233821.1 | - | 5, 6 | 403 |
