siRNA Details
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191578.1 | NP_001178507.1 | 8, 9 | 2392 | ||
XM_006242875.2 | XP_006242937.1 | 11, 12 | 2865 | |||
XM_006242881.2 | XP_006242943.1 | 11, 12 | 2861 | |||
XM_008766141.1 | XP_008764363.1 | 11, 12 | 2862 | |||
GenBank mRNA | X74226.1 | - | 4, 5 | 873 |
