siRNA Details
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191578.1 | NP_001178507.1 | 18 | 3756 | ||
XM_006242875.2 | XP_006242937.1 | 21 | 4229 | |||
XM_006242881.2 | XP_006242943.1 | 20 | 4084 | |||
XM_008766141.1 | XP_008764363.1 | 21 | 4130 | |||
GenBank mRNA | X74226.1 | - | 14 | 2141 |
