siRNA Details
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134401.2 | NP_599228.2 | 1, 2 | 119 | ||
| XM_006231385.2 | XP_006231447.1 | 1, 2 | 181 | |||
| GenBank mRNA | CB692164.1 | - | 1, 2 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crtac1| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134401.2 | NP_599228.2 | 1, 2 | 119 | ||
| XM_006231385.2 | XP_006231447.1 | 1, 2 | 181 | |||
| GenBank mRNA | CB692164.1 | - | 1, 2 | 119 | ||