siRNA Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172017.2 | NP_742014.2 | 5, 6 | 628 | ||
| GenBank mRNA | BC158578.1 | - | 5, 6 | 609 | 7387164 | |
| BG377998.1 | - | 3, 4 | 478 | |||
| CV074812.1 | - | 5, 6 | 608 | |||
| FQ214813.1 | - | 5, 6 | 731 | |||
| FQ219390.1 | - | 5, 6 | 623 | |||
| FQ220202.1 | - | 5, 6 | 625 | |||

Yif1a
Genomic Map