siRNA Details
| Gene Symbol: Marc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134410.2 | NP_599237.1 | 2 | 427 | ||
| GenBank mRNA | AF095741.1 | - | 2 | 428 | ||
| BC061734.1 | - | 2 | 454 | 5598768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marc2| Gene Symbol: Marc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134410.2 | NP_599237.1 | 2 | 427 | ||
| GenBank mRNA | AF095741.1 | - | 2 | 428 | ||
| BC061734.1 | - | 2 | 454 | 5598768 | ||