siRNA Details
| Gene Symbol: Map3k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138503.1 | NP_612512.1 | 6, 7 | 884 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k2| Gene Symbol: Map3k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138503.1 | NP_612512.1 | 6, 7 | 884 | ||