siRNA Details
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342494.7 | XP_342495.4 | 3, 4 | 447 | ||
GenBank mRNA | FN805744.1 | - | 3, 4 | 400 | ||
FQ147184.1 | - | 3, 4 | 409 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342494.7 | XP_342495.4 | 3, 4 | 447 | ||
GenBank mRNA | FN805744.1 | - | 3, 4 | 400 | ||
FQ147184.1 | - | 3, 4 | 409 |