siRNA Details
| Gene Symbol: Pou3f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138837.1 | NP_620192.1 | 1 | 1296 | ||
| GenBank mRNA | AJ001641.1 | - | 1 | 1296 | ||
| M84644.1 | - | 1 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou3f3| Gene Symbol: Pou3f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138837.1 | NP_620192.1 | 1 | 1296 | ||
| GenBank mRNA | AJ001641.1 | - | 1 | 1296 | ||
| M84644.1 | - | 1 | 259 | |||