siRNA Details
| Gene Symbol: Dab2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138710.3 | NP_619724.3 | 7, 8 | 1483 | ||
| XM_006234057.2 | XP_006234119.1 | 6, 7 | 1470 | |||
| XM_008761697.1 | XP_008759919.1 | 4, 5 | 1881 | |||
| XM_008761698.1 | XP_008759920.1 | 4, 5 | 1881 | |||
| XM_008761699.1 | XP_008759921.1 | 4, 5 | 1882 | |||
| GenBank mRNA | BC061865.1 | - | 3, 4 | 797 | 5598046 | |

Dab2ip
Genomic Map