siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138845.2 | NP_620200.2 | 13 | 3792 | ||
| XM_006242061.2 | XP_006242123.1 | 9 | 3901 | |||
| XM_006242063.2 | XP_006242125.1 | 9 | 3900 | |||
| XM_006242064.2 | XP_006242126.1 | 8 | 3741 | |||
| GenBank mRNA | AF275151.1 | - | 5 | 554 | ||

Tnrc6b
Genomic Map