siRNA Details
| Gene Symbol: Gpm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138846.1 | NP_620201.1 | 4 | 721 | ||
| XM_006256849.2 | XP_006256911.1 | 5 | 1260 | |||
| XM_006256850.2 | XP_006256912.1 | 5 | 1262 | |||
| XM_006256851.2 | XP_006256913.1 | 4 | 784 | |||
| XM_008773151.1 | XP_008771373.1 | 4 | 773 | |||
| GenBank mRNA | AB036421.1 | - | 4 | 721 | ||

Gpm6b
Genomic Map