siRNA Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138847.1 | NP_620202.1 | 4 | 478 | ||
| GenBank mRNA | AF158186.1 | - | 4 | 478 | ||
| BC059114.1 | - | 4 | 449 | 6921568 | ||
| BC086589.1 | - | 4 | 401 | 7309991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nip7| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138847.1 | NP_620202.1 | 4 | 478 | ||
| GenBank mRNA | AF158186.1 | - | 4 | 478 | ||
| BC059114.1 | - | 4 | 449 | 6921568 | ||
| BC086589.1 | - | 4 | 401 | 7309991 | ||