siRNA Details
| Gene Symbol: Pvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242963.2 | XP_006243025.1 | 2 | 1159 | ||
| XM_236210.8 | XP_236210.3 | 2 | 1049 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrl1| Gene Symbol: Pvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242963.2 | XP_006243025.1 | 2 | 1159 | ||
| XM_236210.8 | XP_236210.3 | 2 | 1049 | |||