siRNA Details
Gene Symbol: Pvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242963.2 | XP_006243025.1 | 3 | 1517 | ||
XM_236210.8 | XP_236210.3 | 3 | 1407 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242963.2 | XP_006243025.1 | 3 | 1517 | ||
XM_236210.8 | XP_236210.3 | 3 | 1407 |