siRNA Details
| Gene Symbol: Ehd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139324.1 | NP_647540.1 | 3 | 456 | ||
| GenBank mRNA | AY094093.1 | - | 3 | 456 | ||
| BC083175.1 | - | 3 | 438 | 7105816 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehd4| Gene Symbol: Ehd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139324.1 | NP_647540.1 | 3 | 456 | ||
| GenBank mRNA | AY094093.1 | - | 3 | 456 | ||
| BC083175.1 | - | 3 | 438 | 7105816 | ||