siRNA Details
| Gene Symbol: Phf5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138888.1 | NP_620243.1 | 3, 4 | 245 | ||
| GenBank mRNA | AF495522.1 | - | 3, 4 | 245 | ||
| AY321339.1 | - | 261 | ||||
| BC168144.1 | - | 3, 4 | 263 | 8372591 | ||
| FQ223455.1 | - | 1, 2 | 19 | |||

Phf5a
Genomic Map