siRNA Details
| Gene Symbol: Cdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138889.2 | NP_620244.1 | 11 | 1701 | ||
| GenBank mRNA | AF494095.1 | - | 11 | 1698 | ||
| BC085699.1 | - | 11 | 1701 | 7106186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh13| Gene Symbol: Cdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138889.2 | NP_620244.1 | 11 | 1701 | ||
| GenBank mRNA | AF494095.1 | - | 11 | 1698 | ||
| BC085699.1 | - | 11 | 1701 | 7106186 | ||