siRNA Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100640.1 | NP_001094110.1 | 15 | 2434 | ||
| XM_008769602.1 | XP_008767824.1 | 14 | 2396 | |||
| XM_008769603.1 | XP_008767825.1 | 15 | 2272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamc2| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100640.1 | NP_001094110.1 | 15 | 2434 | ||
| XM_008769602.1 | XP_008767824.1 | 14 | 2396 | |||
| XM_008769603.1 | XP_008767825.1 | 15 | 2272 | |||